Thermo Scientific Chemicals

Phenyl acetate, 97%, Thermo Scientific Chemicals

Catalog number: B22561.0B
1000 g, Each
Thermo Scientific Chemicals

Phenyl acetate, 97%, Thermo Scientific Chemicals

Catalog number: B22561.0B
1000 g, Each
Quantity
Catalog number: B22561.0B
also known as B22561-0B
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
122-79-2
IUPAC Name
phenyl acetate
Molecular Formula
C8H8O2
InChI Key
IPBVNPXQWQGGJP-UHFFFAOYSA-N
SMILES
CC(=O)OC1=CC=CC=C1
Appearance (Color)
Clear colorless to pale yellow to pale brown
Form
Liquid
Assay (GC)
≥96.0%
Identification (FTIR)
Conforms
Refractive Index
1.4995-1.5035 @ 20?C

Description

Phenyl acetate levels in urine are marker for the diagnosis of some forms of unipolar major depressive disorders. It undergoes Fries rearrangement to form a mixture of o- and p-hydroxyacetophenones which are useful intermediates in manufacture of pharmaceuticals. It is a metabolite of Phenylbutyrate (PB), useful in the treatment of neuroblastoma and lung cancer. An odorant found in strawberries, passion fruit and black tea.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in water (4 g/L at 20°C).

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text