Assay (Non-aqueous acid-base Titration)≥98.5% to ≤101.5% (dry wt. basis) (non-U.S. specification)
Water Content (Karl Fischer Titration)≤2.0% (U.S. specification)
Optical Rotation-24° ±1° (c=1, water) (non-U.S. specification)
CommentSpecification differs for U.S. and non-U.S. material where indicated
Water Content (Karl Fischer Titration)≤5.0% (non-U.S. specification)
View more