5-Methoxy-2-benzimidazolinone, 98%, Thermo Scientific Chemicals
5-Methoxy-2-benzimidazolinone, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

5-Methoxy-2-benzimidazolinone, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number B22580.03
also known as B22580-03
Price (USD)/ Each
58.10
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
58.10
Add to cart
5-Methoxy-2-benzimidazolinone, 98%, Thermo Scientific Chemicals
Catalog numberB22580.03
Price (USD)/ Each
58.10
-
Add to cart
Chemical Identifiers
CAS2080-75-3
IUPAC Name5-methoxy-2,3-dihydro-1H-1,3-benzodiazol-2-one
Molecular FormulaC8H8N2O2
InChI KeyAUPLVAKFTYFHTA-UHFFFAOYSA-N
SMILESCOC1=CC=C2NC(=O)NC2=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to brown
FormPowder
Assay (HPLC)≥97.5%
Loss on Drying≤2.5% (200?C, const. wt.)
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only