N,N'-Diaminoguanidine monohydrochloride, 98%, Thermo Scientific Chemicals
N,N'-Diaminoguanidine monohydrochloride, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

N,N'-Diaminoguanidine monohydrochloride, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number B22615.14
also known as B22615-14
Price (USD)/ Each
109.00
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
109.00
Add to cart
N,N'-Diaminoguanidine monohydrochloride, 98%, Thermo Scientific Chemicals
Catalog numberB22615.14
Price (USD)/ Each
109.00
-
Add to cart
Chemical Identifiers
CAS36062-19-8
IUPAC Namehydrogen (E)-N,N''-diaminoguanidine chloride
Molecular FormulaCH8ClN5
InChI KeyHAZRIBSLCUYMQP-UHFFFAOYSA-N
SMILES[H+].[Cl-].NN\C(N)=N\N
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormPowder
Assay (Titration ex Chloride)≥97.5 to ≤102.5%
Identification (FTIR)Conforms
Water Content (Karl Fischer Titration)≤1%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only