Thermo Scientific Chemicals

4-Isopropylphenol, 98%, Thermo Scientific Chemicals

Catalog number: B22620.18
50 g, Each
Thermo Scientific Chemicals

4-Isopropylphenol, 98%, Thermo Scientific Chemicals

Catalog number: B22620.18
50 g, Each
Quantity
Catalog number: B22620.18
also known as B22620-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
99-89-8
IUPAC Name
4-(propan-2-yl)phenol
Molecular Formula
C9H12O
InChI Key
YQUQWHNMBPIWGK-UHFFFAOYSA-N
SMILES
CC(C)C1=CC=C(O)C=C1
Assay (GC)
≥97.5%
Identification (FTIR)
Conforms
Melting Point (clear melt)
57-63?C
Appearance (Color)
White or colorless to red/orange or cream to brown
Form
Crystals or powder or crystalline powder or fused/lumpy solid

Description

4-Isopropylphenol was used in the synthesis of vinyl compounds. It is used as a flavor & fragrance agent in cosmetics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Isopropylphenol was used in the synthesis of vinyl compounds. It is used as a flavor & fragrance agent in cosmetics.

Solubility
Slightly soluble in water, soluble in ethanol and ethyl ether.

Notes
Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text