Thermo Scientific Chemicals

Dibenzyl diisopropylphosphoramidite, 90+%, Thermo Scientific Chemicals

Catalog number: B22629.06
5 g, Each
Thermo Scientific Chemicals

Dibenzyl diisopropylphosphoramidite, 90+%, Thermo Scientific Chemicals

Catalog number: B22629.06
5 g, Each
Quantity
Catalog number: B22629.06
also known as B22629-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
108549-23-1
IUPAC Name
[bis(benzyloxy)phosphanyl]bis(propan-2-yl)amine
Molecular Formula
C20H28NO2P
InChI Key
ANPWLBTUUNFQIO-UHFFFAOYSA-N
SMILES
CC(C)N(C(C)C)P(OCC1=CC=CC=C1)OCC1=CC=CC=C1
Appearance (Color)
Clear colorless to pale yellow
Form
Liquid
Assay (HPLC)
≥90.0%
Phosphorus-31 NMR
Conforms to structure
Refractive Index
1.5305-1.5365 @ 20°C

Description

Dibenzyl N,N-diisopropylphosphoramidite may be used for the preparation of phosphopeptides. It is used for the synthesis of a GDP (guanosine diphosphate) analog, SML-8-73-1. It is useful for nucleotide coupling.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dibenzyl N,N-diisopropylphosphoramidite may be used for the preparation of phosphopeptides. It is used for the synthesis of a GDP (guanosine diphosphate) analog, SML-8-73-1. It is useful for nucleotide coupling.

Solubility
Soluble in chloroform.

Notes
Moisture sensitive. Store under inert gas. Store at 0°C. Incompatible with oxidizing agents, water/ moisture.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text