Thermo Scientific Chemicals

1-Iodonaphthalene, 98%, Thermo Scientific Chemicals

Catalog number: B22644.18
50 g, Each
Thermo Scientific Chemicals

1-Iodonaphthalene, 98%, Thermo Scientific Chemicals

Catalog number: B22644.18
50 g, Each
Quantity
Catalog number: B22644.18
also known as B22644-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
90-14-2
IUPAC Name
1-iodonaphthalene
Molecular Formula
C10H7I
InChI Key
NHPPIJMARIVBGU-UHFFFAOYSA-N
SMILES
IC1=C2C=CC=CC2=CC=C1
Assay (GC)
≥97.5%
Appearance (Color)
Clear yellow to brown or purple
Refractive Index
1.6990-1.7030 @ 20°C
Form
Liquid

Description

1-Iodonaphthalene undergoes Pd catalyzed cross-coupling reaction (Stille reaction) with 2,4-dimethoxy-6-(trimethylstannyl)pyrimidine to afford 2,4-dimethoxy-6-(naphthalen-1-yl)pyrimidine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Iodonaphthalene undergoes Pd catalyzed cross-coupling reaction (Stille reaction) with 2,4-dimethoxy-6-(trimethylstannyl)pyrimidine to afford 2,4-dimethoxy-6-(naphthalen-1-yl)pyrimidine.

Solubility
Slightly soluble in water.

Notes
Light sensitive. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text