Thermo Scientific Chemicals

N-Benzyloxycarbonyl-L-serine benzyl ester, 99%, Thermo Scientific Chemicals

Catalog number: B22647.06
5 g, Each
Thermo Scientific Chemicals

N-Benzyloxycarbonyl-L-serine benzyl ester, 99%, Thermo Scientific Chemicals

Catalog number: B22647.06
5 g, Each
Quantity
Catalog number: B22647.06
also known as B22647-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
21209-51-8
IUPAC Name
benzyl 2-{[(benzyloxy)carbonyl]amino}-3-hydroxypropanoate
Molecular Formula
C18H19NO5
InChI Key
MHHDPGHZHFJLBZ-UHFFFAOYNA-N
SMILES
OCC(NC(=O)OCC1=CC=CC=C1)C(=O)OCC1=CC=CC=C1
Form
Powder
Assay (HPLC)
≥98.5%
Melting Point (clear melt)
80.0-86.0?C
Optical Rotation
4.5 ± 1.0? (C=1 in chloroform)
Appearance (Color)
White to cream

Description

N-Benzyloxycarbonyl-L-serine benzyl ester is used as a building block in peptide synthesis and as a biochemical reagent. It is also used to synthesize the corresponding N-benzyloxycarbonyl-O-tert-butyl-(lor dl)-serine benzyl ester. Further, it serves as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Benzyloxycarbonyl-L-serine benzyl ester is used as a building block in peptide synthesis and as a biochemical reagent. It is also used to synthesize the corresponding N-benzyloxycarbonyl-O-tert-butyl-(lor dl)-serine benzyl ester. Further, it serves as a pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text