Methyl 4-hydroxy-3-methoxycinnamate, 99%, Thermo Scientific Chemicals
Methyl 4-hydroxy-3-methoxycinnamate, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Methyl 4-hydroxy-3-methoxycinnamate, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
2 g
Catalog number B22657.09
also known as B22657-09
Price (USD)/ Each
150.65
Online exclusive
167.00 
Save 16.35 (10%)
-
Add to cart
Quantity:
10 g
Request bulk or custom format
Price (USD)/ Each
150.65
Online exclusive
167.00 
Save 16.35 (10%)
Add to cart
Methyl 4-hydroxy-3-methoxycinnamate, 99%, Thermo Scientific Chemicals
Catalog numberB22657.09
Price (USD)/ Each
150.65
Online exclusive
167.00 
Save 16.35 (10%)
-
Add to cart
Chemical Identifiers
CAS2309-07-1
IUPAC Namemethyl (2E)-3-(4-hydroxy-3-methoxyphenyl)prop-2-enoate
Molecular FormulaC11H12O4
InChI KeyAUJXJFHANFIVKH-GQCTYLIASA-N
SMILESCOC(=O)C=CC1=CC=C(O)C(OC)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pink
FormPowder
Assay (GC)≥98.5%
Identification (FTIR)Conforms
Melting Point (clear melt)61.0-68.0?C
Methyl 4-hydroxy-3-methoxycinnamate has antioxidant activities that provide protection against inflammation and cancer. It has got the ability to cross cell membranes. It inhibits the release of pro-inflammatory cytokines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. M.T.Huang; R.C.Smart; C.Q.Wong; A.H.Conney. Inhibitory effect of curcumin, chlorogenic acid, caffeic acid, and ferulic acid on tumor promotion in mouse skin by 12-O-tetradecanoylphorbol-13-acetate. Cancer Res. 1988, 48,(21), 5941-5946.
  2. Rukhsana Sultana. Ferulic acid ethyl ester as a potential therapy in neurodegenerative disorders. Biochimica et Biophysica Acta (BBA) - Molecular Basis of Disease. 2012, 1822,(5), 748-752.