Thermo Scientific Chemicals

2-Diisopropylaminoethyl chloride hydrochloride, 98%, Thermo Scientific Chemicals

Catalog number: B22662.36
500 g, Each
Thermo Scientific Chemicals

2-Diisopropylaminoethyl chloride hydrochloride, 98%, Thermo Scientific Chemicals

Catalog number: B22662.36
500 g, Each
Quantity
Catalog number: B22662.36
also known as B22662-36
Price (USD)
Have Questions?

Chemical Identifiers

CAS
4261-68-1
Assay (Titration ex Chloride)
≥97.5 to ≤102.5%
Identification (FTIR)
Conforms
Water Content (Karl Fischer Titration)
≤1%
Melting Point (clear melt)
130.0-136.0?C
Appearance (Color)
Pale cream to cream

Description

2-Diisopropylaminoethyl chloride hydrochloride is used as an intermediate in pharmaceuticals, agrochemicals, dyestuffs and in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Diisopropylaminoethyl chloride hydrochloride is used as an intermediate in pharmaceuticals, agrochemicals, dyestuffs and in organic synthesis.

Solubility
Soluble in water.

Notes
Hygroscopic. Air sensitive. Incompatible with bases and oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text