Thermo Scientific Chemicals

(+/-)-3-Amino-1,2-propanediol, 97%, Thermo Scientific Chemicals

Catalog number: B22672.36
500 g, Each
Thermo Scientific Chemicals

(+/-)-3-Amino-1,2-propanediol, 97%, Thermo Scientific Chemicals

Catalog number: B22672.36
500 g, Each
Quantity
Catalog number: B22672.36
also known as B22672-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
616-30-8
IUPAC Name
3-aminopropane-1,2-diol
Molecular Formula
C3H9NO2
InChI Key
KQIGMPWTAHJUMN-UHFFFAOYNA-N
SMILES
NCC(O)CO
Appearance (Color)
Clear colorless to yellow
Form
Viscous liquid or low-melting solid
Assay from Supplier's CofA
≥96.0%

Description

3-Amino-1,2-propanediol is used in the preparation of iohexol, which is a X-CT non-ionic contrast medium. It acts as a starting material in the production of specialty materials.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Amino-1,2-propanediol is used in the preparation of iohexol, which is a X-CT non-ionic contrast medium. It acts as a starting material in the production of specialty materials.

Solubility
Miscible with water.

Notes
Hygroscopic. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text