3-Chloro-2-methylpropene, 98%, Thermo Scientific Chemicals
3-Chloro-2-methylpropene, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-Chloro-2-methylpropene, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
50 g
1000 g
Catalog number B22674.30
also known as B22674-30
Price (USD)/ Each
70.65
Online exclusive
83.10 
Save 12.45 (15%)
-
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (USD)/ Each
70.65
Online exclusive
83.10 
Save 12.45 (15%)
Add to cart
3-Chloro-2-methylpropene, 98%, Thermo Scientific Chemicals
Catalog numberB22674.30
Price (USD)/ Each
70.65
Online exclusive
83.10 
Save 12.45 (15%)
-
Add to cart
Chemical Identifiers
CAS563-47-3
IUPAC Name3-chloro-2-methylprop-1-ene
Molecular FormulaC4H7Cl
InChI KeyOHXAOPZTJOUYKM-UHFFFAOYSA-N
SMILESCC(=C)CCl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
Assay (GC)≥97.5% (UK sourced material only)
Assay from Supplier's CofA≥97.5% (GC, US sourced material only)
CommentSourced in UK and US
Identification (FTIR)Conforms (UK sourced material only)
View more
3-Chloro-2-methylpropene has been used as reactant in the synthesis of cyclobutanone. It is used as an intermediate in the production of plastics, pharmaceuticals and other organic chemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Chloro-2-methylpropene has been used as reactant in the synthesis of cyclobutanone. It is used as an intermediate in the production of plastics, pharmaceuticals and other organic chemicals.

Solubility
Not miscible in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

General References:

  1. Po C.Chan; Joseph K.Haseman; Gary A.Boorman; James Huff; Allan G.Manus; Rechard H.Cardy. Forestomach lesions in rats and mice administered 3-chloro-2-methylpropene by gavage for two years. Cancer Res. 1986, 46,6349-6352.
  2. D.A.C.Compton; S.C.Hsi; H.H.Mantsch; W.F.Murphy. The vibrational spectrum, normal coordinate analysis and conformational equilibrium of 3-chloro-2-methylpropene. Journal of Raman Spectroscopy. 1982, 13,(1), 30-37.