Dimethyl suberate, 99%, Thermo Scientific Chemicals
Dimethyl suberate, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Dimethyl suberate, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Catalog number B22681.22
also known as B22681-22
Price (USD)/ Each
124.65
Online exclusive
138.00 
Save 13.35 (10%)
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
124.65
Online exclusive
138.00 
Save 13.35 (10%)
Add to cart
Dimethyl suberate, 99%, Thermo Scientific Chemicals
Catalog numberB22681.22
Price (USD)/ Each
124.65
Online exclusive
138.00 
Save 13.35 (10%)
-
Add to cart
Chemical Identifiers
CAS1732-09-8
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Assay (GC)≥98.5%
Refractive Index1.4325-1.4355 @ 20?C
FormLiquid
Dimethyl suberate was used in the preparation of [8-2H2] and [7-2H2]oleates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dimethyl suberate was used in the preparation of [8-2H2] and [7-2H2]oleates.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. D.H.Nugteren. Arachidonate lipoxygenase in blood platelets. Biochimica et Biophysica Acta (BBA) - Lipids and Lipid Metabolism. 1975, 380,(2), 299-307.
  2. A.P.Tulloch. Synthesis of [11-2H2], [8-2H2], [7-2H2], [6-2H2], [5-2H2], [4-2H2] and [3-2H2] cis-9-octadecenoates. Chemistry and Physics of Lipids. 1979, 25,(3), 225-235.