Thermo Scientific Chemicals

4-n-Dodecyloxybenzoic acid, 98%, Thermo Scientific Chemicals

Catalog number: B22689.06
5 g, Each
Thermo Scientific Chemicals

4-n-Dodecyloxybenzoic acid, 98%, Thermo Scientific Chemicals

Catalog number: B22689.06
5 g, Each
Quantity
Catalog number: B22689.06
also known as B22689-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2312-15-4
IUPAC Name
4-(dodecyloxy)benzoic acid
Molecular Formula
C19H30O3
InChI Key
ALQLYJHDBAKLBB-UHFFFAOYSA-N
SMILES
CCCCCCCCCCCCOC1=CC=C(C=C1)C(O)=O
Appearance (Color)
White
Assay (Aqueous acid-base Titration)
≥97.5%
Form
Powder

Description

4-n-Dodecyloxybenzoic acid is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-n-Dodecyloxybenzoic acid is used as an organic chemical synthesis intermediate.

Solubility
Insoluble in water. Soluble in N,N-Dimethylformamide.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text