Assay (Aqueous acid-base Titration)≥98.0 to ≤102.0%
FormPowder
Appearance (Color)White
Optical Rotation26.1 ± 0.1? (C=2 in acetic acid)
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.