4-Methoxy-4'-nitrobenzophenone, 97%, Thermo Scientific Chemicals
4-Methoxy-4'-nitrobenzophenone, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Methoxy-4'-nitrobenzophenone, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
25 g
Catalog number B22720.06
also known as B22720-06
Price (USD)/ Each
142.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
142.00
Add to cart
4-Methoxy-4'-nitrobenzophenone, 97%, Thermo Scientific Chemicals
Catalog numberB22720.06
Price (USD)/ Each
142.00
-
Add to cart
Chemical Identifiers
CAS1151-94-6
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream to pale yellow
Assay (GC)≥96.0%
FormPowder
Melting Point (clear melt)119.0-128.0?C
4-Methoxy-4'-nitrobenzophenone is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Methoxy-4′-nitrobenzophenone is used as an organic chemical synthesis intermediate.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. Hiromu Terao; Yuzo Itoh; Kayo Ohno; Masato Isogai; Atsushi Kakuta; Akio Mukoh. Second-order nonlinear optical properties and polymorphism of benzophenone derivatives. Optics Communications. 1990, 75,(5-6), 451-453.
  2. Mitsuru Shindo; Yusuke Sato; Kozo Shishido. Stereoelectronic effect on stereoselective olefination of ketones providing tetrasubstituted olefins via ynolates. J. Org. Chem. 2000, 65,(17), 5443-5445.