Thermo Scientific Chemicals

4-Hydroxypiperidine, 97%, Thermo Scientific Chemicals

Catalog number: B22723.22
100 g, Each
Thermo Scientific Chemicals

4-Hydroxypiperidine, 97%, Thermo Scientific Chemicals

Catalog number: B22723.22
100 g, Each
Quantity
Catalog number: B22723.22
also known as B22723-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
5382-16-1
IUPAC Name
4-hydroxypiperidin-1-ium
Molecular Formula
C5H12NO
InChI Key
HDOWRFHMPULYOA-UHFFFAOYSA-O
SMILES
OC1CC[NH2+]CC1
Form
Crystalline powder or crystals and/or chunks
Water Content (Karl Fischer Titration)
<1.5%
Appearance (Color)
White to cream or pale yellow to yellow
Melting Point
83-92?C
Assay (GC)
≥96.0%

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text