3,4-Dimethylanisole, 99%, Thermo Scientific Chemicals
3,4-Dimethylanisole, 99%, Thermo Scientific Chemicals
Thermo Scientific™

3,4-Dimethylanisole, 99%, Thermo Scientific Chemicals

 
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
50 g
Catalog number B22735.30
also known as B22735-30
Price (USD)/ Each
216.00
-
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (USD)/ Each
216.00
Add to cart
3,4-Dimethylanisole, 99%, Thermo Scientific Chemicals
Catalog numberB22735.30
Price (USD)/ Each
216.00
-
Add to cart
Chemical Identifiers
CAS4685-47-6
IUPAC Name4-methoxy-1,2-dimethylbenzene
Molecular FormulaC9H12O
InChI KeyLVUBSVWMOWKPDJ-UHFFFAOYSA-N
SMILESCOC1=CC=C(C)C(C)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
Identification (FTIR)Conforms
FormLiquid
Assay (GC)≥98.5%
Refractive Index1.5160-1.5190 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
 
RUO – Research Use Only