Thermo Scientific Chemicals

5-Methoxyisatin, 97%, Thermo Scientific Chemicals

Catalog number: B22742.06
5 g, Each
Thermo Scientific Chemicals

5-Methoxyisatin, 97%, Thermo Scientific Chemicals

Catalog number: B22742.06
5 g, Each
Quantity
Catalog number: B22742.06
also known as B22742-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
39755-95-8
IUPAC Name
5-methoxy-2,3-dihydro-1H-indole-2,3-dione
Molecular Formula
C9H7NO3
InChI Key
DMHGXMPXHPOXBF-UHFFFAOYSA-N
SMILES
COC1=CC=C2NC(=O)C(=O)C2=C1
Appearance (Color)
Dark red to brown or dark red/brown
Assay (HPLC)
≥96.0%
Form
Powder
Melting Point (clear melt)
188-206°C

Description

5-Methoxyisatin is used for Pharmaceutical intermediates, a raw material used in chemical synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Methoxyisatin is used for Pharmaceutical intermediates, a raw material used in chemical synthesis.

Solubility
Soluble in cold water.

Notes
Store in cool, dry conditions in a sealed container. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text