5-Methoxyisatin is used for Pharmaceutical intermediates, a raw material used in chemical synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
5-Methoxyisatin is used for Pharmaceutical intermediates, a raw material used in chemical synthesis.
Solubility
Soluble in cold water.
Notes
Store in cool, dry conditions in a sealed container. Incompatible with oxidizing agents.
RUO – Research Use Only
G. Bryant Bachman; George M. Picha. Synthesis of Substituted Aminobenzacridines. J. Am. Chem. Soc. 1946, 68 (8), 1599-1602.
Paul G. Gassman; Berkeley W. CueJr; Tien-Yau Luh. A general method for the synthesis of isatins. J. Org. Chem. 1977, 42 (8), 1344-1348.