Thermo Scientific Chemicals

1,2,3,4-Tetrahydroquinoline, 98%, Thermo Scientific Chemicals

Catalog number: B22750.22
100 g, Each
Thermo Scientific Chemicals

1,2,3,4-Tetrahydroquinoline, 98%, Thermo Scientific Chemicals

Catalog number: B22750.22
100 g, Each
Quantity
Catalog number: B22750.22
also known as B22750-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
635-46-1
IUPAC Name
1,2,3,4-tetrahydroquinoline
Molecular Formula
C9H11N
InChI Key
LBUJPTNKIBCYBY-UHFFFAOYSA-N
SMILES
C1CNC2=CC=CC=C2C1
Appearance (Color)
Clear pale yellow to yellow to brown
Assay (GC)
≥97.5%
Refractive Index
1.5930-1.5970 @ 20?C
Identification (FTIR)
Conforms
Form
Liquid

Description

1,2,3,4-Tetrahydroquinoline is used as an antioxidant and corrosion inhibitor. It is an active component in various dyes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,2,3,4-Tetrahydroquinoline is used as an antioxidant and corrosion inhibitor. It is an active component in various dyes.

Solubility
Miscible with alcohol, ether, carbon disulfide and many organic solvents. Slightly miscible with water.

Notes
Hygroscopic. Incomaptible with strong oxidizing agents.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text