Thermo Scientific Chemicals

3-Chloro-4-fluorobenzeneboronic acid, 98%, Thermo Scientific Chemicals

Catalog number: B22755.14
25 g, Each
Thermo Scientific Chemicals

3-Chloro-4-fluorobenzeneboronic acid, 98%, Thermo Scientific Chemicals

Catalog number: B22755.14
25 g, Each
Quantity
Catalog number: B22755.14
also known as B22755-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
144432-85-9
IUPAC Name
(3-chloro-4-fluorophenyl)boronic acid
Molecular Formula
C6H5BClFO2
InChI Key
WJDZZXIDQYKVDG-UHFFFAOYSA-N
SMILES
OB(O)C1=CC=C(F)C(Cl)=C1
Proton NMR
Conforms to structure
Form
Powder
Assay (HPLC)
≥97.5%
Assay (Aqueous acid-base Titration)
≥97.5%
Appearance (Color)
White to pale cream

Description

3-Chloro-4-fluorobenzeneboronic acid is used as a reactant for Rh-catalyzed asymmetric addition reactions, palladium-catalyzed oxidative cross-coupling reaction and Suzuki-Miyaura coupling.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Chloro-4-fluorobenzeneboronic acid is used as a reactant for Rh-catalyzed asymmetric addition reactions, palladium-catalyzed oxidative cross-coupling reaction and Suzuki-Miyaura coupling.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text