Thermo Scientific Chemicals

2-Aminofluorene, 98%, Thermo Scientific Chemicals

Catalog number: B22769.14
25 g, Each
Thermo Scientific Chemicals

2-Aminofluorene, 98%, Thermo Scientific Chemicals

Catalog number: B22769.14
25 g, Each
Quantity
Catalog number: B22769.14
also known as B22769-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
153-78-6
IUPAC Name
9H-fluoren-2-amine
Molecular Formula
C13H11N
InChI Key
CFRFHWQYWJMEJN-UHFFFAOYSA-N
SMILES
NC1=CC=C2C(CC3=CC=CC=C23)=C1
Appearance (Color)
White to cream to yellow to pale brown
Form
Powder or crystalline powder
Assay (GC)
≥97.0%
Melting Point (clear melt)
123-132°C
Identification (FTIR)
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text