Thermo Scientific Chemicals

1-Chloro-3,4-dinitrobenzene, tech. 90%, Thermo Scientific Chemicals

Catalog number: B22773.22
100 g, Each
Thermo Scientific Chemicals

1-Chloro-3,4-dinitrobenzene, tech. 90%, Thermo Scientific Chemicals

Catalog number: B22773.22
100 g, Each
Quantity
Catalog number: B22773.22
also known as B22773-22
Price (EUR)
Quantity
-

Chemical Identifiers

CAS
610-40-2
IUPAC Name
4-chloro-1,2-dinitrobenzene
Molecular Formula
C6H3ClN2O4
InChI Key
QVQSOXMXXFZAKU-UHFFFAOYSA-N
SMILES
[O-][N+](=O)C1=CC=C(Cl)C=C1[N+]([O-])=O
Appearance (Color)
Clear yellow to brown
Form
Fused solid or liquid as melt
Assay (GC)
≥88.0%
Refractive Index
1.5815-1.5895 @ 20?C

Description

1-Chloro-3,4-dinitrobenzene is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Chloro-3,4-dinitrobenzene is used as pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text

    Safety and Handling


    Classification of the substance or mixture
    CLP classification - Regulation(EC) No 1272/2008
    Acute oral toxicity
    Category 3
    Acute dermal toxicity
    Category 3
    Acute Inhalation Toxicity - Vapors
    Category 2
    Specific target organ toxicity - (repeated exposure)
    Category 2
    Acute aquatic toxicity
    Category 1
    Chronic aquatic toxicity
    Category 1
    Label Elements
    Signal Word

    Danger

    Hazard Statements

    H301 + H311 - Toxic if swallowed or in contact with skin

    H330 - Fatal if inhaled

    H373 - May cause damage to organs through prolonged or repeated exposure

    H410 - Very toxic to aquatic life with long lasting effects

    Precautionary Statements

    P280 - Wear protective gloves/protective clothing

    P302 + P352 - IF ON SKIN: Wash with plenty of soap and water

    P304 + P340 - IF INHALED: Remove person to fresh air and keep comfortable for breathing

    P310 - Immediately call a POISON CENTER or doctor/physician