Thermo Scientific Chemicals

1-Chloro-3,4-dinitrobenzene, tech. 90%, Thermo Scientific Chemicals

Catalog number: B22773.30
250 g, Each
Thermo Scientific Chemicals

1-Chloro-3,4-dinitrobenzene, tech. 90%, Thermo Scientific Chemicals

Catalog number: B22773.30
250 g, Each
Quantity
Catalog number: B22773.30
also known as B22773-30
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
610-40-2
IUPAC Name
4-chloro-1,2-dinitrobenzene
Molecular Formula
C6H3ClN2O4
InChI Key
QVQSOXMXXFZAKU-UHFFFAOYSA-N
SMILES
[O-][N+](=O)C1=CC=C(Cl)C=C1[N+]([O-])=O
Appearance (Color)
Clear yellow to brown
Form
Fused solid or liquid as melt
Assay (GC)
≥88.0%
Refractive Index
1.5815-1.5895 @ 20?C

Description

1-Chloro-3,4-dinitrobenzene is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Chloro-3,4-dinitrobenzene is used as pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text