Thermo Scientific Chemicals

3,5-Diamino-1,2,4-triazole, 98%, Thermo Scientific Chemicals

Catalog number: B22775.22
100 g, Each
Thermo Scientific Chemicals

3,5-Diamino-1,2,4-triazole, 98%, Thermo Scientific Chemicals

Catalog number: B22775.22
100 g, Each
Quantity
Catalog number: B22775.22
also known as B22775-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1455-77-2
IUPAC Name
1H-1,2,4-triazole-3,5-diamine
Molecular Formula
C2H5N5
InChI Key
PKWIYNIDEDLDCJ-UHFFFAOYSA-N
SMILES
NC1=NC(N)=NN1
Identification (FTIR)
Conforms
Appearance (Color)
White to cream to pale brown or grey
Assay (Non-aqueous acid-base Titration)
≥97.5 to ≤102.5%
Form
Crystals or powder or crystalline powder

Description

3,5-Diamino-1,2,4-triazole is used as an inhibitor of DNA synthesis. It also serves as an antitumor agents in the treatment of epigenetically-based diseases. It acts as a corrosion inhibitor for copper.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in water.

Notes
Air sensitive. Incompatible with acids and oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text