4-Phenyl-2-butanone, 98%, Thermo Scientific Chemicals
4-Phenyl-2-butanone, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Phenyl-2-butanone, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1000 g
Catalog number B22777.30
also known as B22777-30
Price (USD)/ Each
63.00
-
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (USD)/ Each
63.00
Add to cart
4-Phenyl-2-butanone, 98%, Thermo Scientific Chemicals
Catalog numberB22777.30
Price (USD)/ Each
63.00
-
Add to cart
Chemical Identifiers
CAS2550-26-7
IUPAC Name4-phenylbutan-2-one
Molecular FormulaC10H12O
InChI KeyAKGGYBADQZYZPD-UHFFFAOYSA-N
SMILESCC(=O)CCC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
FormLiquid
Assay (GC)≥97.5%
Identification (FTIR)Conforms
Refractive Index1.5105-1.5145 @ 20?C
4-Phenyl-2-butanone be used as an attractant for melon flies and as an odorant for soap. It is also used in the preparation of 4-oxocyclohexanecarbaldehyde derivatives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Phenyl-2-butanone be used as an attractant for melon flies and as an odorant for soap. It is also used in the preparation of 4-oxocyclohexanecarbaldehyde derivatives.

Solubility
Not miscible in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. Melvin S.Newman; Gideon Fraenkel; Walter N.Kirn. The reactions of 4-phenyl-2-butanone and 5-phenyl-2-pentanone with phosphorus pentachloride1. J. Org. Chem. 1963, 28,(7), 1851-1853.
  2. John B.Melpolder; Richard F.Heck. Palladium-catalyzed arylation of allylic alcohols with aryl halides. J. Org. Chem. 1976, 41,(2), 265-272.