Thermo Scientific Chemicals

2,3-Dichlorobenzeneboronic acid, 98%, Thermo Scientific Chemicals

Catalog number: B22781.03
1 g, Each
Thermo Scientific Chemicals

2,3-Dichlorobenzeneboronic acid, 98%, Thermo Scientific Chemicals

Catalog number: B22781.03
1 g, Each
Quantity
Catalog number: B22781.03
also known as B22781-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
151169-74-3
IUPAC Name
(2,3-dichlorophenyl)boronic acid
Molecular Formula
C6H5BCl2O2
InChI Key
TYIKXPOMOYDGCS-UHFFFAOYSA-N
SMILES
OB(O)C1=C(Cl)C(Cl)=CC=C1
Appearance (Color)
White to cream
Assay (HPLC)
≥97.5%
Assay (Aqueous acid-base Titration)
≥97.5%
Form
Crystals or powder or crystalline powder
Proton NMR
Conforms to structure

Description

2,3-Dichlorobenzeneboronic acid is used as a reactant for Suzuki-Miyaura coupling reactions. It is also used in preparation of biologically and pharmacologically active molecules.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,3-Dichlorobenzeneboronic acid is used as a reactant for Suzuki-Miyaura coupling reactions. It is also used in preparation of biologically and pharmacologically active molecules.

Solubility
Soluble in methanol.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text