2,4,5-Trimethoxybenzaldehyde, 98%, Thermo Scientific Chemicals
2,4,5-Trimethoxybenzaldehyde, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2,4,5-Trimethoxybenzaldehyde, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number B22792.22
also known as B22792-22
Price (USD)/ Each
172.00
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
172.00
Add to cart
2,4,5-Trimethoxybenzaldehyde, 98%, Thermo Scientific Chemicals
Catalog numberB22792.22
Price (USD)/ Each
172.00
-
Add to cart
Chemical Identifiers
CAS4460-86-0
IUPAC Name2,4,5-trimethoxybenzaldehyde
Molecular FormulaC10H12O4
InChI KeyIAJBQAYHSQIQRE-UHFFFAOYSA-N
SMILESCOC1=CC(OC)=C(C=O)C=C1OC
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥97.5%
Identification (FTIR)Conforms
Free acid (titration)≤1.0%
Melting Point111.0-115.5?C
Appearance (Color)White to cream
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only