4-Amino-2,2-difluoro-1,3-benzodioxole, 97+%, Thermo Scientific Chemicals
4-Amino-2,2-difluoro-1,3-benzodioxole, 97+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Amino-2,2-difluoro-1,3-benzodioxole, 97+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number B22797.06
also known as B22797-06
Price (USD)/ Each
392.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
392.00
Add to cart
4-Amino-2,2-difluoro-1,3-benzodioxole, 97+%, Thermo Scientific Chemicals
Catalog numberB22797.06
Price (USD)/ Each
392.00
-
Add to cart
Specifications
Chemical Name or Material4-Amino-2,2-difluoro-1,3-benzodioxole
CAS106876-54-4
Health Hazard 1H301-H312+H332-H315-H319
Health Hazard 2GHS H Statement
H301-H312-H332-H315-H319
Toxic if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
View more
4-Amino-2,2-difluoro-1,3-benzodioxole is used in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Amino-2,2-difluoro-1,3-benzodioxole is used in organic synthesis.

Solubility
Not miscible or difficult to mix in water.

Notes
Store in cool, dry conditions in a sealed container. Incompatible with oxidizing agents.
RUO – Research Use Only