3,4-Difluorobenzeneboronic acid, 97%, Thermo Scientific Chemicals
3,4-Difluorobenzeneboronic acid, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3,4-Difluorobenzeneboronic acid, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number B22799.03
also known as B22799-03
Price (USD)/ Each
33.50
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
33.50
Add to cart
3,4-Difluorobenzeneboronic acid, 97%, Thermo Scientific Chemicals
Catalog numberB22799.03
Price (USD)/ Each
33.50
-
Add to cart
Chemical Identifiers
CAS168267-41-2
IUPAC Name(3,4-difluorophenyl)boronic acid
Molecular FormulaC6H5BF2O2
InChI KeyRMGYQBHKEWWTOY-UHFFFAOYSA-N
SMILESOB(O)C1=CC=C(F)C(F)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Assay (HPLC)≥96.0%
FormCrystals or powder or crystalline powder
Proton NMRConforms to structure
Assay (Aqueous acid-base Titration)≥96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only