Thermo Scientific Chemicals

2,6-Difluorobenzeneboronic acid, 98%, Thermo Scientific Chemicals

Catalog number: B22805.03
1 g, Each
Thermo Scientific Chemicals

2,6-Difluorobenzeneboronic acid, 98%, Thermo Scientific Chemicals

Catalog number: B22805.03
1 g, Each
Quantity
Catalog number: B22805.03
also known as B22805-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
162101-25-9
IUPAC Name
(2,6-difluorophenyl)boronic acid
Molecular Formula
C6H5BF2O2
InChI Key
DBZAICSEFBVFHL-UHFFFAOYSA-N
SMILES
OB(O)C1=C(F)C=CC=C1F
Form
Crystals or powder or crystalline powder
Assay (HPLC)
≥97.5%
Appearance (Color)
White to pale cream
Assay (Aqueous acid-base Titration)
≥97.5%
Proton NMR
Conforms to structure

Description

2,6-Difluorobenzeneboronic acid is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,6-Difluorobenzeneboronic acid is used as an organic chemical synthesis intermediate.

Solubility
Soluble in methanol.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text