Benzene-1,3-disulfonic acid disodium salt is used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Benzene-1,3-disulfonic acid disodium salt is used as pharmaceutical intermediate.
Solubility
Soluble in water( 663 g/L at 20°C).
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only
Maocheng Yan; Dennis E.Tallman; Gordon P.Bierwagen. Role of oxygen in the galvanic interaction between polypyrrole and aluminum alloy. Electrochimica Acta. 2008, 54,(2), 220-227.
C.Sarzanini; M.C.Bruzzoniti; G.Sacchero; E.Mentasti. Retention model for anionic, neutral, and cationic analytes in reversed-phase ion interaction chromatography. Anal. Chem. 1996, 68,(24), 4494-4500.