Benzene-1,3-disulfonic acid disodium salt, 94%, Thermo Scientific Chemicals
Benzene-1,3-disulfonic acid disodium salt, 94%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Benzene-1,3-disulfonic acid disodium salt, 94%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
100 g
Catalog number B22811.36
also known as B22811-36
Price (USD)/ Each
208.00
-
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (USD)/ Each
208.00
Add to cart
Benzene-1,3-disulfonic acid disodium salt, 94%, Thermo Scientific Chemicals
Catalog numberB22811.36
Price (USD)/ Each
208.00
-
Add to cart
Chemical Identifiers
CAS831-59-4
IUPAC Namedisodium benzene-1,3-disulfonate
Molecular FormulaC6H4Na2O6S2
InChI KeyXWPWZOJBTOJEGW-UHFFFAOYSA-L
SMILES[Na+].[Na+].[O-]S(=O)(=O)C1=CC(=CC=C1)S([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream
Assay (ex Sulphated Ash)≥92.0%
FormPowder
Benzene-1,3-disulfonic acid disodium salt is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Benzene-1,3-disulfonic acid disodium salt is used as pharmaceutical intermediate.

Solubility
Soluble in water( 663 g/L at 20°C).

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. Maocheng Yan; Dennis E.Tallman; Gordon P.Bierwagen. Role of oxygen in the galvanic interaction between polypyrrole and aluminum alloy. Electrochimica Acta. 2008, 54,(2), 220-227.
  2. C.Sarzanini; M.C.Bruzzoniti; G.Sacchero; E.Mentasti. Retention model for anionic, neutral, and cationic analytes in reversed-phase ion interaction chromatography. Anal. Chem. 1996, 68,(24), 4494-4500.