Thermo Scientific Chemicals

Trimethylhydroquinone, 98%, Thermo Scientific Chemicals

Catalog number: B22819.22
100 g, Each
Thermo Scientific Chemicals

Trimethylhydroquinone, 98%, Thermo Scientific Chemicals

Catalog number: B22819.22
100 g, Each
Quantity
Catalog number: B22819.22
also known as B22819-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
700-13-0
IUPAC Name
2,3,5-trimethylbenzene-1,4-diol
Molecular Formula
C9H12O2
InChI Key
AUFZRCJENRSRLY-UHFFFAOYSA-N
SMILES
CC1=CC(O)=C(C)C(C)=C1O
Form
Powder
Melting Point (clear melt)
167.0-173.0?C
Appearance (Color)
White to cream
Assay (GC)
≥97.5%

Description

Trimethylhydroquinone is used for synthesizing vitamin E and used as medical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Trimethylhydroquinone is used for synthesizing vitamin E and used as medical intermediate.

Solubility
Soluble in water (2 g/L @ 20°C.

Notes
Store in cool, dry conditions in a sealed container. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text