Thermo Scientific Chemicals

Benzo[b]thiophene-2-boronic acid, 97%, Thermo Scientific Chemicals

Catalog number: B22835.14
25 g, Each
Thermo Scientific Chemicals

Benzo[b]thiophene-2-boronic acid, 97%, Thermo Scientific Chemicals

Catalog number: B22835.14
25 g, Each
Quantity
Catalog number: B22835.14
also known as B22835-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
98437-23-1
IUPAC Name
(1-benzothiophen-2-yl)boronic acid
Molecular Formula
C8H7BO2S
InChI Key
YNCYPMUJDDXIRH-UHFFFAOYSA-N
SMILES
OB(O)C1=CC2=CC=CC=C2S1
Appearance (Color)
White to cream
Assay (Aqueous acid-base Titration)
≥96.0%
Form
Powder
Proton NMR
Conforms to structure

Description

Benzo[b]thiophene-2-boronic acid is used in the synthesis of phosphorescent sensor for quantification of copper(II) ion, UV promoted phenanthridine syntheses, Preparation of CYP11B1 inhibitors for treatment of cortisol dependent diseases, PDE4 inhibitors, Suzuki-Miyaura cross-coupling reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Insoluble in water.

Notes
Store in cool, dry conditions in a sealed container. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text