Thermo Scientific Chemicals

(Di-n-dodecyl)dimethylammonium bromide, 98+%, Thermo Scientific Chemicals

Catalog number: B22839.04
2 g, Each
Thermo Scientific Chemicals

(Di-n-dodecyl)dimethylammonium bromide, 98+%, Thermo Scientific Chemicals

Catalog number: B22839.04
2 g, Each
Quantity
Catalog number: B22839.04
also known as B22839-04
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
3282-73-3
IUPAC Name
didodecyldimethylazanium bromide
Molecular Formula
C26H56BrN
InChI Key
XRWMGCFJVKDVMD-UHFFFAOYSA-M
SMILES
[Br-].CCCCCCCCCCCC[N+](C)(C)CCCCCCCCCCCC
Appearance (Color)
White to pale cream
Melting Point (clear melt)
155.0-166.0°C
Assay (Titration ex Bromide)
≥98.0 to ≤102.0%
Loss on Drying
≤1.0%
Form
Powder or crystals

Description

(Di-n-dodecyl)dimethylammonium bromide is used as a phase transfer catalyst. It is also used as floor disinfectant. Further, it acts as surface-active agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(Di-n-dodecyl)dimethylammonium bromide is used as a phase transfer catalyst. It is also used as floor disinfectant. Further, it acts as surface-active agent.

Solubility
Soluble in hot and cold water.

Notes
Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text