Thermo Scientific Chemicals

Phenyltrimethylammonium chloride, 98+%, Thermo Scientific Chemicals

Catalog number: B22851.36
500 g, Each
Thermo Scientific Chemicals

Phenyltrimethylammonium chloride, 98+%, Thermo Scientific Chemicals

Catalog number: B22851.36
500 g, Each
Quantity
Catalog number: B22851.36
also known as B22851-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
138-24-9
IUPAC Name
N,N,N-trimethylanilinium chloride
Molecular Formula
C9H14ClN
InChI Key
MQAYPFVXSPHGJM-UHFFFAOYSA-M
SMILES
[Cl-].C[N+](C)(C)C1=CC=CC=C1
Appearance (Color)
White
Assay (Titration ex Chloride)
≥98.0 to ≤102.0%
Form
Crystals or powder or crystalline powder
Water Content (Karl Fischer Titration)
≤1.0%

Description

Phenyltrimethylammonium chloride is an effective phase transfer catalyst, methylating agent, also useful for alkaline depolymerizations, corrosion inhibitor for low carbon steel.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Phenyltrimethylammonium chloride is an effective phase transfer catalyst, methylating agent, also useful for alkaline depolymerizations, corrosion inhibitor for low carbon steel.

Solubility
Soluble in water 333 g/L.

Notes
Hygroscopic. Store under inert gas. Store away from oxidizing agent, water/ moisture.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text