N-[Tris(hydroxymethyl)methyl] acrylamide is used to prepare N-(5-hydroxymethyl-2-phenyl-[1,3]dioxan-5-yl)-acrylamide by reaction with benzaldehyde.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
N-[Tris(hydroxymethyl)methyl] acrylamide is used to prepare N-(5-hydroxymethyl-2-phenyl-[1,3]dioxan-5-yl)-acrylamide by reaction with benzaldehyde.
Solubility
Soluble in water.
Notes
Hygroscopic. Light sensitive. Incompatible with strong oxdizing agents.
RUO – Research Use Only
Liao, Y.; An, N.; Wang, N.; Zhang, Y.; Song, J.; Zhou, J.; Liu, W. Photoactive Self-Shaping Hydrogels as Noncontact 3D Macro/Microscopic Photoprinting Platforms. Macromol. Rapid Commun. 2015, 36 (24), 2129-2136.
Kotanen, C. N.; Janagam, D. R.; Idziak, R.; Rhym, L.; Sullivan, R.; Wilson, A. M.; Lowe, T. L.; Guiseppi-Elie, A. Partitioning of coomassie brilliant blue into DMAEMA containing poly(HEMA)-based hydrogels. Eur. Polym. J. 2015, 72, 438-450.