N-[Tris(hydroxymethyl)methyl]acrylamide, 99%, Thermo Scientific Chemicals
N-[Tris(hydroxymethyl)methyl]acrylamide, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

N-[Tris(hydroxymethyl)methyl]acrylamide, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
100 g
Catalog number B22854.14
also known as B22854-14
Price (USD)/ Each
323.65
Online exclusive
359.00 
Save 35.35 (10%)
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
323.65
Online exclusive
359.00 
Save 35.35 (10%)
Add to cart
N-[Tris(hydroxymethyl)methyl]acrylamide, 99%, Thermo Scientific Chemicals
Catalog numberB22854.14
Price (USD)/ Each
323.65
Online exclusive
359.00 
Save 35.35 (10%)
-
Add to cart
Chemical Identifiers
CAS13880-05-2
IUPAC NameN-[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]prop-2-enamide
Molecular FormulaC7H13NO4
InChI KeyMVBJSQCJPSRKSW-UHFFFAOYSA-N
SMILESOCC(CO)(CO)NC(=O)C=C
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Assay (Titration ex Chloride)≤0.1%
Appearance (Color)White
Assay (HPLC)≥98.5%
N-[Tris(hydroxymethyl)methyl] acrylamide is used to prepare N-(5-hydroxymethyl-2-phenyl-[1,3]dioxan-5-yl)-acrylamide by reaction with benzaldehyde.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-[Tris(hydroxymethyl)methyl] acrylamide is used to prepare N-(5-hydroxymethyl-2-phenyl-[1,3]dioxan-5-yl)-acrylamide by reaction with benzaldehyde.

Solubility
Soluble in water.

Notes
Hygroscopic. Light sensitive. Incompatible with strong oxdizing agents.
RUO – Research Use Only

General References:

  1. Liao, Y.; An, N.; Wang, N.; Zhang, Y.; Song, J.; Zhou, J.; Liu, W. Photoactive Self-Shaping Hydrogels as Noncontact 3D Macro/Microscopic Photoprinting Platforms. Macromol. Rapid Commun. 2015, 36 (24), 2129-2136.
  2. Kotanen, C. N.; Janagam, D. R.; Idziak, R.; Rhym, L.; Sullivan, R.; Wilson, A. M.; Lowe, T. L.; Guiseppi-Elie, A. Partitioning of coomassie brilliant blue into DMAEMA containing poly(HEMA)-based hydrogels. Eur. Polym. J. 2015, 72, 438-450.