Thermo Scientific Chemicals

3-Bromo-1-propanol, 95%, Thermo Scientific Chemicals

Catalog number: B22857.14
25 g, Each
Thermo Scientific Chemicals

3-Bromo-1-propanol, 95%, Thermo Scientific Chemicals

Catalog number: B22857.14
25 g, Each
Quantity
Catalog number: B22857.14
also known as B22857-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
627-18-9
IUPAC Name
3-bromopropan-1-ol
Molecular Formula
C3H7BrO
InChI Key
RQFUZUMFPRMVDX-UHFFFAOYSA-N
SMILES
OCCCBr
Form
Liquid
Appearance (Color)
Clear colorless to pale yellow
Assay (GC)
≥94.0
Identification (FTIR)
Conforms
Refractive Index
1.4865-1.4925 @ 20°C

Description

3-Bromo-1-propanol is used in the synthesis of fluorescent halide-sensitive quinolinium dyes, chiral, quaternary prolines through cyclization of quaternary amino acids and molten salt-polymers. It is utilized for the study of micellar media and in microemulsions based on cationic or a nonionic surfactant by reacting with phenols. It acts as a grafting agent in the synthesis of recyclable reagents and an intermediate in the synthesis of organic molecules.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Bromo-1-propanol is used in the synthesis of fluorescent halide-sensitive quinolinium dyes, chiral, quaternary prolines through cyclization of quaternary amino acids and molten salt-polymers. It is utilized for the study of micellar media and in microemulsions based on cationic or a nonionic surfactant by reacting with phenols. It acts as a grafting agent in the synthesis of recyclable reagents and an intermediate in the synthesis of organic molecules.

Solubility
Miscible with water. Slightly miscible with ethanol and ether.

Notes
Store in cool place. Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text