Thermo Scientific Chemicals

4-Chlorobenzyl bromide, 98+%, Thermo Scientific Chemicals

Catalog number: B22858.14
25 g, Each
Thermo Scientific Chemicals

4-Chlorobenzyl bromide, 98+%, Thermo Scientific Chemicals

Catalog number: B22858.14
25 g, Each
Quantity
Catalog number: B22858.14
also known as B22858-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
622-95-7
IUPAC Name
1-(bromomethyl)-4-chlorobenzene
Molecular Formula
C7H6BrCl
InChI Key
KQNBRMUBPRGXSL-UHFFFAOYSA-N
SMILES
ClC1=CC=C(CBr)C=C1
Appearance (Color)
White
Assay (GC)
>98.0%
Form
crystalline powder or fused solid

Description

4-Chlorobenzyl bromide is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Chlorobenzyl bromide is used as pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents, bases and active metals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text