Thermo Scientific Chemicals

(+/-)-2-Aminoheptane, 98+%, Thermo Scientific Chemicals

Catalog number: B22863.14
25 g, Each
Thermo Scientific Chemicals

(+/-)-2-Aminoheptane, 98+%, Thermo Scientific Chemicals

Catalog number: B22863.14
25 g, Each
Quantity
Catalog number: B22863.14
also known as B22863-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
123-82-0
IUPAC Name
heptan-2-amine
Molecular Formula
C7H17N
InChI Key
VSRBKQFNFZQRBM-UHFFFAOYNA-N
SMILES
CCCCCC(C)N
Refractive Index
1.4170-1.4210 @20?C
Identification (FTIR)
Conforms
Appearance (Color)
Clear colorless to yellow
Assay (GC)
≥98.0%
Form
Liquid

Description

(±)-2-Aminoheptane is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(+/-)-2-Aminoheptane is used as an organic chemical synthesis intermediate.

Solubility
Soluble in water ( 9 g/L at 20°C).

Notes
Air sensitive. Incompatible with oxidizing agents and carbon dioxide.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text