Thermo Scientific Chemicals

1-Naphthyl phosphate disodium salt hydrate, 99%, Thermo Scientific Chemicals

Catalog number: B22866.14
25 g, Each
Thermo Scientific Chemicals

1-Naphthyl phosphate disodium salt hydrate, 99%, Thermo Scientific Chemicals

Catalog number: B22866.14
25 g, Each
Quantity
Catalog number: B22866.14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
207569-06-0
IUPAC Name
disodium naphthalen-1-yl phosphate
Molecular Formula
C10H7Na2O4P
InChI Key
QYURIFWAOPAPAJ-UHFFFAOYSA-L
SMILES
[Na+].[Na+].[O-]P([O-])(=O)OC1=C2C=CC=CC2=CC=C1
Water Content (Karl Fischer Titration)
3.10-9.40% (0.5-1.5 waters)
Identification (FTIR)
Conforms
Appearance (Color)
White to cream or pale pink or pale brown
Form
Powder
Assay (Non-aqueous acid-base Titration)
≥98.5 to ≤101.5% (dry wt. basis)

Description

1-Naphthyl phosphate disodium salt hydrate is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Naphthyl phosphate disodium salt hydrate is used as an organic chemical synthesis intermediate.

Solubility
Soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and heat.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text