Thermo Scientific Chemicals

1-Bromo-2,4-dinitrobenzene, 98%, Thermo Scientific Chemicals

Catalog number: B22867.22
100 g, Each
Thermo Scientific Chemicals

1-Bromo-2,4-dinitrobenzene, 98%, Thermo Scientific Chemicals

Catalog number: B22867.22
100 g, Each
Quantity
Catalog number: B22867.22
also known as B22867-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
584-48-5
IUPAC Name
1-bromo-2,4-dinitrobenzene
Molecular Formula
C6H3BrN2O4
InChI Key
PBOPJYORIDJAFE-UHFFFAOYSA-N
SMILES
[O-][N+](=O)C1=CC=C(Br)C(=C1)[N+]([O-])=O
Appearance (Color)
Pale cream to cream to yellow
Assay (GC)
≥97.5%
Melting Point (clear melt)
68.0-75.0?C
Form
Crystals or powder or crystalline powder

Description

1-Bromo-2,4-dinitrobenzene has been used in the preparation of 2,4-dinitrophenol via treatment with KO2-crown ether complex in benzene. It is used as substrate in protein determination and glutathione S-transferase (GST) assay of chicken and rat prostaglandin D2 synthase (PGDS).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Bromo-2,4-dinitrobenzene has been used in the preparation of 2,4-dinitrophenol via treatment with KO2-crown ether complex in benzene. It is used as substrate in protein determination and glutathione S-transferase (GST) assay of chicken and rat prostaglandin D2 synthase (PGDS).

Solubility
Soluble in water (partly miscible).

Notes
Store in cool, dry conditions in a sealed container. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text