Thermo Scientific Chemicals

1,4-Diisopropylbenzene, 98%, Thermo Scientific Chemicals

Catalog number: B22871.36
500 g, Each
Thermo Scientific Chemicals

1,4-Diisopropylbenzene, 98%, Thermo Scientific Chemicals

Catalog number: B22871.36
500 g, Each
Quantity
Catalog number: B22871.36
also known as B22871-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
100-18-5
IUPAC Name
1,4-bis(propan-2-yl)benzene
Molecular Formula
C12H18
InChI Key
SPPWGCYEYAMHDT-UHFFFAOYSA-N
SMILES
CC(C)C1=CC=C(C=C1)C(C)C
Assay (GC)
≥98.5%
Form
Liquid
Appearance (Color)
Clear colorless
Refractive Index
1.4875-1.4920 @ 20?C

Description

1,4-Diisopropylbenzene is used as an intermediate for the synthesis of stabilizers, polymers, synthetic lubricants and hydroperoxides. It is also used in the trivalent actinide-lanthanide separation by phosphorus reagent extraction from aqueous komplexes (TALSPEAK) process for separating trivalent lanthanides from trivalent actinide cations. It finds application in nuclear fuel reprocessing.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,4-Diisopropylbenzene is used as an intermediate for the synthesis of stabilizers, polymers, synthetic lubricants and hydroperoxides. It is also used in the trivalent actinide-lanthanide separation by phosphorus reagent extraction from aqueous komplexes (TALSPEAK) process for separating trivalent lanthanides from trivalent actinide cations. It finds application in nuclear fuel reprocessing.

Solubility
Soluble in alcohol, ether, acetone and benzene. Insoluble in water.

Notes
Incompatible with oxidizing agents and strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text