Thermo Scientific Chemicals

1,12-Dodecanediol, 99%, Thermo Scientific Chemicals

Catalog number: B22885.14
25 g, Each
Thermo Scientific Chemicals

1,12-Dodecanediol, 99%, Thermo Scientific Chemicals

Catalog number: B22885.14
25 g, Each
Quantity
Catalog number: B22885.14
also known as B22885-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
5675-51-4
IUPAC Name
dodecane-1,12-diol
Molecular Formula
C12H26O2
InChI Key
GHLKSLMMWAKNBM-UHFFFAOYSA-N
SMILES
OCCCCCCCCCCCCO
Appearance (Color)
White
Form
Crystals or powder or crystalline powder or flakes or pellets
Melting Point (clear melt)
78.0-84.0?C
Assay (Silylated GC)
≥98.5%

Description

1,12-Dodecanediol is used in the polyester polyols for polyurethanes, coatings and inks, adhesives, lastomers, polyesters and co polyesters, polymer cross linkers, pharmaceutical intermediates, fragrances.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,12-Dodecanediol is used in the polyester polyols for polyurethanes, coatings and inks, adhesives, lastomers, polyesters and co polyesters, polymer cross linkers, pharmaceutical intermediates, fragrances.

Solubility
Soluble in alcohol and warm ether. Insoluble in water and petroleum ether.

Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text