1-Aminoanthraquinone, 97%, Thermo Scientific Chemicals
1-Aminoanthraquinone, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1-Aminoanthraquinone, 97%, Thermo Scientific Chemicals

Have Questions?
Quantity:
100 g
Catalog number B22901.22
also known as B22901-22
Price (USD)/ Each
43.00
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
43.00
Add to cart
1-Aminoanthraquinone, 97%, Thermo Scientific Chemicals
Catalog numberB22901.22
Price (USD)/ Each
43.00
-
Add to cart
Chemical Identifiers
CAS82-45-1
IUPAC Name1-amino-9,10-dihydroanthracene-9,10-dione
Molecular FormulaC14H9NO2
InChI KeyKHUFHLFHOQVFGB-UHFFFAOYSA-N
SMILESNC1=CC=CC2=C1C(=O)C1=CC=CC=C1C2=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Identification (FTIR)Conforms
Appearance (Color)Dark red to red-brown
Assay (HPLC)≥96.0%
Melting Point250.0-255.0?C
In the manufacturing of dyes and pharmaceuticals

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only