Di-n-pentyl phthalate, 98%, Thermo Scientific Chemicals
Di-n-pentyl phthalate, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Di-n-pentyl phthalate, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number B22910.06
also known as B22910-06
Price (USD)/ Each
99.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
117.00 
Save 17.35 (15%)
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
99.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
117.00 
Save 17.35 (15%)
Add to cart
Di-n-pentyl phthalate, 98%, Thermo Scientific Chemicals
Catalog numberB22910.06
Price (USD)/ Each
99.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
117.00 
Save 17.35 (15%)
-
Add to cart
Chemical Identifiers
CAS131-18-0
IUPAC Name1,2-dipentyl benzene-1,2-dicarboxylate
Molecular FormulaC18H26O4
InChI KeyIPKKHRVROFYTEK-UHFFFAOYSA-N
SMILESCCCCCOC(=O)C1=CC=CC=C1C(=O)OCCCCC
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥97.5%
Refractive Index1.4875-1.4915 @ 20?C
Appearance (Color)Clear colorless to pale yellow
FormLiquid
Di-n-pentyl phthalate is a plasticizer for PVC membrane electrodes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Di-n-pentyl phthalate is a plasticizer for PVC membrane electrodes.

Solubility
Not miscible or difficult to mix in water.

Notes
Store in cool, dry conditions in a sealed container. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. Dianne M. Creasy; John R. Foster and Paul M. D. Foster. The morphological development of di-n-pentyl phthalate induced testicular atrophy in the rat. The Journal of Pathology. 1983, 139 (3), 309-321.
  2. Paul M.D. Foster; John R. Foster, Melvyn W. Cook; Lucy V. Thomas; Sharat D. Changes in ultrastructure and cytochemical localization of zinc in rat testis following the administration of Di-n-pentyl phthalate. Toxicology and Applied Pharmacology. 1982, 63, (1), 120-132.