Thermo Scientific Chemicals

Di-n-pentyl phthalate, 98%, Thermo Scientific Chemicals

Catalog number: B22910.22
100 g, Each
Thermo Scientific Chemicals

Di-n-pentyl phthalate, 98%, Thermo Scientific Chemicals

Catalog number: B22910.22
100 g, Each
Quantity
Catalog number: B22910.22
also known as B22910-22
Price (USD)
Have Questions?

Chemical Identifiers

CAS
131-18-0
IUPAC Name
1,2-dipentyl benzene-1,2-dicarboxylate
Molecular Formula
C18H26O4
InChI Key
IPKKHRVROFYTEK-UHFFFAOYSA-N
SMILES
CCCCCOC(=O)C1=CC=CC=C1C(=O)OCCCCC
Assay (GC)
≥97.5%
Refractive Index
1.4875-1.4915 @ 20?C
Appearance (Color)
Clear colorless to pale yellow
Form
Liquid

Description

Di-n-pentyl phthalate is a plasticizer for PVC membrane electrodes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Di-n-pentyl phthalate is a plasticizer for PVC membrane electrodes.

Solubility
Not miscible or difficult to mix in water.

Notes
Store in cool, dry conditions in a sealed container. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text